Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Bis(2-chloroethoxy)ethane 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR54670-100G OR54670-25G OR54670-500G
77410-416EA 44.28 USD InStock
77410-416 77410-418 77410-420
1,2-Bis(2-chloroethoxy)ethane 97%
1,2-Bis(2-chloroethoxy)ethane


Formula: ClH₂CCH₂OCH₂CH₂OCH₂CH₂Cl
MW: 187.07 g/mol
Boiling Pt: 235
Melting Pt: -31.5
Density: 1.197 g/cm³ (25 °C)
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00000976
CAS Number: 112-26-5
UN: 2810
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty