Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Diisocyanatobutane

Supplier: APOLLO SCIENTIFIC
Warning

OR55710-10G OR55710-1G OR55710-5G
77410-510EA 126.34 USD InStock
77410-510 77410-512 77410-514
1,4-Diisocyanatobutane
1,4-Diisocyanatobutane
  • May form small amount of sediment.


Formula: C₆H₈N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00075559
CAS Number: 4538-37-8
UN: 3334
ADR: 9,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty