Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-iodo-1H-indazole-5-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

OR60118-100MG OR60118-250MG
77410-672EA 21.34 USD InStock
77410-672 77410-674
Methyl-3-iodo-1H-indazole-5-carboxylate
Methyl 3-iodo-1H-indazole-5-carboxylate


Formula: C₉H₇IN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06739144
CAS Number: 885271-25-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty