Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Bromomethyl)-2-methoxybenzene, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

OR350580-1G OR350580-250MG OR350580-5G
77409-298EA 296.48 USD InStock
77409-298 77409-300 77409-302
1-(Bromomethyl)-2-methoxybenzene, Technical Grade
1-(Bromomethyl)-2-methoxybenzene


Formula: C₈H₉BrO
Boiling Pt: 118 °C (18 mmHg)
Melting Pt: 47…50 °C
Storage Temperature: Freezer
MDL Number: MFCD09745128
CAS Number: 52289-93-7
UN: 1759
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty