Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxypyrazine 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR40046-1G OR40046-5G
77409-462EA 21.26 USD InStock
77409-462 77409-466
2-Hydroxypyrazine 95%
2-Hydroxypyrazine


Formula: C₄H₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD00233966
CAS Number: 6270-63-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty