Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Collidine

Supplier: APOLLO SCIENTIFIC
Danger

OR4784-100ML OR4784-500ML
77409-992EA 83.54 USD InStock
77409-992 77409-994
2,4,6-Collidine
2,4,6-Collidine


Formula: C₈H₁₁N
Boiling Pt: 171…172 °C
Melting Pt: –43 °C
Density: 0.917 g/cm³ (25 °C)
Flash Pt: 55 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006338
CAS Number: 108-75-8
EINECS: 203-613-3
UN: 1992
ADR: 3+6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty