Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Dodecylbenzenesulfonic acid

Supplier: APOLLO SCIENTIFIC
Danger

OR52171-100G OR52171-25G
77410-222EA 66.22 USD InStock
77410-222 77410-224
4-Dodecylbenzenesulfonic acid
4-Dodecylbenzenesulfonic acid


Formula: C₁₈H₃₀O₃S
MW: 326.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066677
CAS Number: 121-65-3
UN: 2586
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty