Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Isocyanato-1,3-benzodioxole 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR61306-1G OR61306-5G
77410-724EA 52.98 USD InStock
77410-724 77410-726
5-Isocyanato-1,3-benzodioxole 95%
5-Isocyanato-1,3-benzodioxole
When melted, may contain small amount of sediment.


Formula: C₈H₅NO₃
Melting Pt: 48…51 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01863692
CAS Number: 69922-28-7
UN: 3335
ADR: 9,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty