Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dichloro-2-propanol 95%

Supplier: APOLLO SCIENTIFIC

OR61346-100G OR61346-25G
77410-734EA 39.74 USD InStock
77410-734 77410-736
1,3-Dichloro-2-propanol 95%
1,3-Dichloro-2-propanol


Formula: ClH₂CCHOHCH₂Cl
Boiling Pt: 174…176 °C
Melting Pt: 4 °C
Density: 1.351 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00000951
CAS Number: 96-23-1
UN: 2750
ADR: 6.1,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty