Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bromoacetamide 96%

Supplier: APOLLO SCIENTIFIC
Danger

OR6592-25G OR6592-5G
77410-876EA 66.22 USD InStock
77410-876 77410-878
Bromoacetamide 96%
Bromoacetamide


Formula: BrCH₂CONH₂
Melting Pt: 87…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00008025
CAS Number: 683-57-8
UN: 2923
ADR: 8+6.1,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty