Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methoxy-2-naphthaldehyde 98%

Supplier: APOLLO SCIENTIFIC

OR900156-100G OR900156-25G
77411-486EA 166.06 USD InStock
77411-486 77411-488
6-Methoxy-2-naphthaldehyde 98%
6-Methoxy-2-naphthaldehyde


Formula: C₁₂H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00081152
CAS Number: 3453-33-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty