Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Prop-2-yn-1-yl)morpholine 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR48271-1G OR48271-25G OR48271-5G
77410-046EA 21.32 USD InStock
77410-046 77410-048 77410-050
4-(Prop-2-yn-1-yl)morpholine 98%
4-(Prop-2-yn-1-yl)morpholine


Formula: C₇H₁₁NO
Boiling Pt: 186 °C
Storage Temperature: Ambient
MDL Number: MFCD00510919
CAS Number: 5799-76-8
UN: 1993
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty