Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Thienylacetic acid 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR5060-25G OR5060-5G
77410-120EA 39.74 USD InStock
77410-120 77410-122
3-Thienylacetic acid 98%
3-Thienylacetic acid


Formula: C₆H₆O₂S
Storage Temperature: Ambient
MDL Number: MFCD00005473
CAS Number: 6964-21-2
EINECS: 230-166-1
UN: 3261
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty