Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,5-Dimethylbicyclo[4.1.0]heptan-2-one

Supplier: APOLLO SCIENTIFIC

OR50906-1G OR50906-250MG OR50906-5G
77410-128EA 484.96 USD InStock
77410-128 77410-130 77410-132
5,5-Dimethylbicyclo[4.1.0]heptan-2-one
5,5-Dimethylbicyclo[4.1.0]heptan-2-one


Formula: C₉H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD24691569
CAS Number: 88105-44-6
UN: 1993
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty