Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Acetyl-2-chloropyridine 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR61610-1G OR61610-5G
77410-776EA 21.26 USD InStock
77410-776 77410-778
3-Acetyl-2-chloropyridine 97%
3-Acetyl-2-chloropyridine


Formula: C₇H₆ClNO
MW: 155.5816 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03840751
CAS Number: 55676-21-6
UN: 2810
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty