Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-(trifluoromethyl)anisole 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC0384-10G PC0384-1G PC0384-5G
77412-530EA 158.86 USD InStock
77412-530 77412-532 77412-534
3-Fluoro-5-(trifluoromethyl)anisole 95%
3-Fluoro-5-(trifluoromethyl)anisole


Formula: C₈H₆F₄O
Flash Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD01631491
CAS Number: 261951-79-5
UN: 1993
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty