Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-ethylbenzenesulfonyl chloride 95+%

Supplier: APOLLO SCIENTIFIC
Danger

OR5613-1G OR5613-250MG OR5613-5G
77410-526EA 74.38 USD InStock
77410-526 77410-528 77410-530
4-Bromo-2-ethylbenzenesulfonyl chloride 95+%
4-Bromo-2-ethylbenzenesulfonyl chloride


Formula: C₈H₈BrClO₂S
Boiling Pt: 101…104 °C/0.3 mmHg
Melting Pt: 43…45 °C
Storage Temperature: Ambient
MDL Number: MFCD00204178
CAS Number: 175278-24-7
UN: 3261
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty