Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-isocyanato-1-methylbenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR59455-1G OR59455-25G OR59455-5G
77410-596EA 22.42 USD InStock
77410-596 77410-598 77410-600
4-Chloro-2-isocyanato-1-methylbenzene 98%
4-Chloro-2-isocyanato-1-methylbenzene
  • May form small amount of sediment.


Formula: C₈H₆ClNO
MW: 167.6 g/mol
Boiling Pt: 115 °C/5 mmHg
Density: 1.226 g/cm³ (25 °C)
Flash Pt: 108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00019913
CAS Number: 40411-27-6
UN: 2206
ADR: 6.1 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty