Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-1-methyl-1H-indole 95+%

Supplier: APOLLO SCIENTIFIC
Warning

OR9630-1G OR9630-250MG OR9630-5G
77412-248EA 83.64 USD InStock
77412-248 77412-250 77412-252
6-Bromo-1-methyl-1H-indole 95+%
6-Bromo-1-methyl-1H-indole


Formula: C₉H₈BrN
MW: 210.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08543444
CAS Number: 125872-95-9
UN: 2810
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty