Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dihydro-5-[(methylamino)methyl]thieno[3,4-b][1,4]dioxine 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR9137-1G OR9137-250MG
77411-692EA 186.44 USD InStock
77411-692 77411-694
2,3-Dihydro-5-[(methylamino)methyl]thieno[3,4-b][1,4]dioxine 95%
2,3-Dihydro-5-[(methylamino)methyl]thieno[3,4-b][1,4]dioxine


Formula: C₈H₁₁NO₂S
Storage Temperature: Ambient
MDL Number: MFCD08271957
CAS Number: 859851-03-9
UN: 2922
ADR: 8+6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty