Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Butyl methanesulfonate 95%

Supplier: APOLLO SCIENTIFIC

OR928832-10G OR928832-25G OR928832-5G
77411-920EA 79.46 USD InStock
77411-920 77411-922 77411-924
Butyl methanesulfonate 95%
Butyl methanesulfonate


Formula: C₅H₁₂O₃S
Storage Temperature: Ambient
MDL Number: MFCD00041257
CAS Number: 1912-32-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty