Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4,5,5-Tetramethyl-1,3,2-dioxaborolane

Supplier: APOLLO SCIENTIFIC
Danger

OR7123-100G OR7123-25G OR7123-5G
77411-046EA 137.54 USD InStock
77411-046 77411-048 77411-050
4,4,5,5-Tetramethyl-1,3,2-dioxaborolane
4,4,5,5-Tetramethyl-1,3,2-dioxaborolane
Reportedly couples with aryl halides to produce aryl boronates:J.Org.Chem.(1997)62,6458-6459.


Formula: C₆H₁₃BO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00674030
CAS Number: 25015-63-8
UN: 3399
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty