Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dibromobutanoyl chloride, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

OR938772-1G OR938772-25G OR938772-5G
77412-052EA 75.4 USD InStock
77412-052 77412-054 77412-056
2,4-Dibromobutanoyl chloride, Technical Grade
2,4-Dibromobutanoyl chloride


Formula: C₄H₅Br₂ClO
MW: 264.35 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01862994
CAS Number: 82820-87-9
UN: 3265
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty