Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,6-Dimethyl-4-octyne-3,6-diol 95%

Supplier: APOLLO SCIENTIFIC

OR950160-25G OR950160-500G
77412-148EA 110.04 USD InStock
77412-148 77412-150
3,6-Dimethyl-4-octyne-3,6-diol 95%
3,6-Dimethyl-4-octyne-3,6-diol


Formula: C₁₀H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD00004480
CAS Number: 78-66-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty