Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Chloro-6-fluorobenzylthio)ethylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC1864Q-1G PC1864Q-250MG PC1864Q-5G
77413-394EA 103.92 USD InStock
77413-394 77413-396 77413-398
2-(2-Chloro-6-fluorobenzylthio)ethylamine 97%
2-(2-Chloro-6-fluorobenzylthio)ethylamine


Formula: C₉H₁₁ClFNS
Boiling Pt: 140 °C/6 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00052683
CAS Number: 175136-76-2
UN: 3261
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty