Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DL-Methionine methyl ester hydrochloride 95%

Supplier: APOLLO SCIENTIFIC

OR916765-25G OR916765-5G
77411-706EA 69.28 USD InStock
77411-706 77411-708
DL-Methionine methyl ester hydrochloride 95%
DL-Methionine methyl ester hydrochloride


Formula: C₆H₁₃NO₂S·HCl
MW: 199.7 g/mol
Melting Pt: 110…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00067540
CAS Number: 16118-36-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty