Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-fluoro-3-methylbenzoyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

PC0100-1G PC0100-25G PC0100-5G
77412-388EA 38.72 USD InStock
77412-388 77412-390 77412-392
2-Chloro-6-fluoro-3-methylbenzoyl chloride
2-Chloro-6-fluoro-3-methylbenzoyl chloride


Formula: C₈H₅Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD01631371
CAS Number: 261762-82-7
UN: 3265
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty