Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc formate 95+%

Supplier: APOLLO SCIENTIFIC
Warning

OR922222-100G OR922222-25G OR922222-500G
77411-782EA 100.86 USD InStock
77411-782 77411-784 77411-786
Zinc formate 95+%
Zinc formate


Formula: C₂H₂O₄Zn
MW: 155.40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00050836
CAS Number: 557-41-5
UN: 3077
ADR: 9+EHS III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty