Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(1H-1,2,4-Triazol-1-yl)benzaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR9235-1G OR9235-250MG
77411-864EA 70.3 USD InStock
77411-864 77411-866
3-(1H-1,2,4-Triazol-1-yl)benzaldehyde 97%
3-(1H-1,2,4-Triazol-1-yl)benzaldehyde


Formula: C₉H₇N₃O
MW: 173.1714 g/mol
Melting Pt: 114.5…115.5 °C
Storage Temperature: Ambient
MDL Number: MFCD08271896
CAS Number: 868755-54-8
UN: 1325
ADR: 4.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty