Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Fur-2-yl)benzyl alcohol 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR9307-1G OR9307-250MG OR9307-5G
77411-948EA 140.6 USD InStock
77411-948 77411-950 77411-952
2-(Fur-2-yl)benzyl alcohol 97%
2-(Fur-2-yl)benzyl alcohol


Formula: C₁₁H₁₀O₂
Melting Pt: 85…86.5 °C
Storage Temperature: Ambient
MDL Number: MFCD06203101
CAS Number: 139697-88-4
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty