Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-4-hydroxy-2-(trifluoromethyl)quinoline 95+%

Supplier: APOLLO SCIENTIFIC
Danger

PC1476-1G PC1476-5G
77413-194EA 73.4 USD InStock
77413-194 77413-196
6-Bromo-4-hydroxy-2-(trifluoromethyl)quinoline 95+%
6-Bromo-4-hydroxy-2-(trifluoromethyl)quinoline


Formula: C₁₀H₅BrF₃NO
MW: 292.06 g/mol
Melting Pt: >290 °C
Storage Temperature: Ambient
MDL Number: MFCD00153078
CAS Number: 1701-22-0
UN: 2811
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty