Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4,5-difluorobenzonitrile 98%

Supplier: APOLLO SCIENTIFIC

PC1504-25G PC1504-50G PC1504-5G
77413-202EA 195.62 USD InStock
77413-202 77413-204 77413-206
2-Chloro-4,5-difluorobenzonitrile 98%
2-Chloro-4,5-difluorobenzonitrile


Formula: C₇H₂ClF₂N
Melting Pt: 33…36 °C
Storage Temperature: Ambient
MDL Number: MFCD02871619
CAS Number: 135748-34-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty