Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluorobiphenyl 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC1423I-100G PC1423I-25G
77413-170EA 38.72 USD InStock
77413-170 77413-172
4-Bromo-2-fluorobiphenyl 97%
4-Bromo-2-fluorobiphenyl


Formula: C₁₂H₈BrF
MW: 251.1 g/mol
Boiling Pt: 299
Melting Pt: 38.5
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00051716
CAS Number: 41604-19-7
EINECS: 255-453-9
UN: 3152
ADR: 9 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty