Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-fluoropyridine 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC2546-100G PC2546-25G PC2546-5G
77413-678EA 158.94 USD InStock
77413-678 77413-680 77413-682
2-Chloro-5-fluoropyridine 99%
2-Chloro-5-fluoropyridine


Formula: C₅H₃ClFN
Boiling Pt: 74 °C/38 mmHg
Melting Pt: 19 °C
Density: 1.297 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD03095332
CAS Number: 31301-51-6
UN: 1993
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty