Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chlorofluoroacetic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC1148-1G PC1148-5G
77413-062EA 93.74 USD InStock
77413-062 77413-064
Chlorofluoroacetic acid 97%
Chlorofluoroacetic acid


Formula: C₂H₂ClFO₂
MW: 112.49 g/mol
Boiling Pt: 162 °C
Density: 1.532 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00871521
CAS Number: 471-44-3
UN: 3265
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty