Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Fluoro-o-toluidine 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC1391-100G PC1391-25G PC1391-5G
77413-140EA 154.86 USD InStock
77413-140 77413-142 77413-144
6-Fluoro-o-toluidine 97%
6-Fluoro-o-toluidine


Formula: C₇H₈FN
MW: 125.1435 g/mol
Boiling Pt: 88…89 °C/13 mmHg
Density: 1.082 g/cm³ (25 °C)
Flash Pt: 77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06658252
CAS Number: 443-89-0
UN: 2811
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty