Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-[(Trifluoromethyl)thio]benzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC1572-100G PC1572-25G
77413-218EA 825.24 USD InStock
77413-218 77413-220
4-[(Trifluoromethyl)thio]benzonitrile 98%
4-[(Trifluoromethyl)thio]benzonitrile


Formula: C₈H₄F₃NS
MW: 203.19 g/mol
Melting Pt: 39…43 °C
Storage Temperature: Ambient
MDL Number: MFCD00039475
CAS Number: 332-26-3
UN: 2811
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty