Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-1,1,2-trifluoro-1-methoxyethane 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC2070-100G PC2070-25G
77413-500EA 636.4 USD InStock
77413-500 77413-502
2-Chloro-1,1,2-trifluoro-1-methoxyethane 97%
2-Chloro-1,1,2-trifluoro-1-methoxyethane


Formula: C₃H₄ClF₃O
Boiling Pt: 69…71.5 °C
Melting Pt: –92 °C
Density: 1.363 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00039335
CAS Number: 425-87-6
EINECS: 207-038-9
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty