Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-methyl-3-(trifluoromethyl)benzaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC303145-1G PC303145-5G
77414-364EA 320.92 USD InStock
77414-364 77414-366
2-Fluoro-5-methyl-3-(trifluoromethyl)benzaldehyde 98%
2-Fluoro-5-methyl-3-(trifluoromethyl)benzaldehyde


Formula: C₉H₆F₄O
Storage Temperature: Ambient
MDL Number: MFCD28054185
CAS Number: 1706458-33-4
UN: 2811
ADR: 6.1,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty