Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Fluoro-2-iodoethane 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC0560-1G PC0560-25G PC0560-5G
77412-630EA 21.4 USD InStock
77412-630 77412-632 77412-634
1-Fluoro-2-iodoethane 98%
1-Fluoro-2-iodoethane


Formula: C₂H₄FI
MW: 173.96 g/mol
Boiling Pt: 98…102 °C
Density: 2.136 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00748131
CAS Number: 762-51-6
UN: 2810
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty