Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluorocatechol 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC0672-1G PC0672-500MG
77412-674EA 117.16 USD InStock
77412-674 77412-676
4-Fluorocatechol 97%
4-Fluorocatechol


Formula: C₆H₅FO₂
MW: 128.10 g/mol
Melting Pt: 90…91 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01317539
CAS Number: 367-32-8
EINECS: 226-356-9
UN: 1759
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty