Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-3-methylbenzo[b]thiophene-2-sulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

PC2907-1G PC2907-5G
77413-972EA 166.06 USD InStock
77413-972 77413-974
5-Fluoro-3-methylbenzo[b]thiophene-2-sulfonyl chloride
5-Fluoro-3-methylbenzo[b]thiophene-2-sulfonyl chloride


Formula: C₉H₆ClFO₂S₂
MW: 264.724 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02091512
CAS Number: 404964-34-7
UN: 3261
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty