Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-2-fluorosulfur pentafluoride 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC303266-1G PC303266-250MG
77414-378EA 1730.96 USD InStock
77414-378 77414-380
4-Amino-2-fluorosulfur pentafluoride 98%
4-Amino-2-fluorosulfur pentafluoride


Formula: C₆H₅F₆NS
Storage Temperature: Ambient
MDL Number: MFCD28054193
CAS Number: 864230-06-8
UN: 2811
ADR: 6.1,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty