Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-2-methylbenzotrifluoride 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC3319-1G PC3319-25G PC3319-5G
77414-694EA 21.26 USD InStock
77414-694 77414-696 77414-698
3-Fluoro-2-methylbenzotrifluoride 98%
3-Fluoro-2-methylbenzotrifluoride


Formula: C₈H₆F₄
MW: 178.13 g/mol
Density: 1.2 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD08062608
CAS Number: 910911-43-2
UN: 1993
ADR: 3 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty