Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ammonium tetrafluoroborate 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC1110-100G PC1110-25G PC1110-500G
77413-006EA 23.4 USD InStock
77413-006 77413-008 77413-010
Ammonium tetrafluoroborate 98%
Ammonium tetrafluoroborate


Formula: H₄BF₄N
Melting Pt: 230 °C
Density: 1.871 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD06245785
CAS Number: 13826-83-0
EINECS: 237-531-4
UN: 3260
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty