Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-o-toluonitrile

Supplier: APOLLO SCIENTIFIC
Warning

PC1841-100G PC1841-25G PC1841-5G
77413-336EA 92.72 USD InStock
77413-336 77413-338 77413-340
4-Fluoro-o-toluonitrile
4-Fluoro-o-toluonitrile


Formula: C₈H₆FN
MW: 135.14
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD03095106
CAS Number: 147754-12-9
UN: 3439
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty