Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-fluoro-6-nitroaniline 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC3591-1G PC3591-5G
77414-774EA 21.26 USD InStock
77414-774 77414-776
2-Bromo-4-fluoro-6-nitroaniline 97%
2-Bromo-4-fluoro-6-nitroaniline


Formula: C₆H₄BrFN₂O₂
MW: 235.01 g/mol
Melting Pt: 74…75 °C
Storage Temperature: Ambient
MDL Number: MFCD07779528
CAS Number: 10472-88-5
UN: 2811
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty