Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-(trifluoromethyl)pyridine 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC2437-100G PC2437-25G PC2437-5G
77413-644EA 279.16 USD InStock
77413-644 77413-646 77413-648
2-Chloro-4-(trifluoromethyl)pyridine 98%
2-Chloro-4-(trifluoromethyl)pyridine


Formula: C₆H₃ClF₃N
MW: 181.54 g/mol
Boiling Pt: 145…147 °C
Melting Pt: -1
Density: 1.411 g/cm³ (25 °C)
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00042224
CAS Number: 81565-18-6
UN: 1992
ADR: 3+6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty