Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluorophenol 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC2865-25G PC2865-5G
77413-928EA 37.7 USD InStock
77413-928 77413-930
2,3-Difluorophenol 98%
2,3-Difluorophenol


Formula: C₆H₄F₂O
Boiling Pt: 53…55 °C/25 mmHg
Melting Pt: 34…38 °C
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00010262
CAS Number: 6418-38-8
UN: 1325
ADR: 4.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty