Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-2-fluoro-3-methoxybenzoyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC302416-1G PC302416-25G PC302416-5G
77414-170EA 61.12 USD InStock
77414-170 77414-172 77414-174
6-Chloro-2-fluoro-3-methoxybenzoyl chloride 97%
6-Chloro-2-fluoro-3-methoxybenzoyl chloride


Formula: C₈H₅Cl₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD04115948
CAS Number: 886499-63-4
UN: 3265
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty